Assay Details
| Gene Symbol: PAOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718768.3 | XP_002718814.3 | 3 - 4 | 1029 | 60 | |
| XM_017348630.1 | XP_017204119.1 | 3 - 4 | 1333 | 60 | ||
More Information
Set Membership: |
|

PAOX
Genomic Map