Assay Details
Gene Symbol: SUGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712973.3 | XP_002713019.1 | 6 - 7 | 576 | 64 | |
XM_008260023.2 | XP_008258245.1 | 6 - 7 | 587 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
