Assay Details
| Gene Symbol: C9H3orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343817.1 | XP_017199306.1 | 2 - 3 | 631 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9H3orf62| Gene Symbol: C9H3orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017343817.1 | XP_017199306.1 | 2 - 3 | 631 | 115 | |
Set Membership: |
|