Assay Details
Gene Symbol: LOC100346409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008275329.2 | XP_008273551.1 | 1127 | 71 | ||
XM_008275330.2 | XP_008273552.1 | 936 | 71 | |||
XM_008275331.2 | XP_008273553.1 | 1127 | 71 | |||
XM_017337556.1 | XP_017193045.1 | 1127 | 71 | |||
XM_017337557.1 | XP_017193046.1 | 668 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
