Assay Details
| Gene Symbol: LEMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268512.2 | XP_008266734.1 | 3 - 4 | 331 | 72 | |
| XM_008268514.2 | XP_008266736.1 | 3 - 4 | 331 | 72 | ||
| XM_008268515.2 | XP_008266737.1 | 3 - 4 | 331 | 72 | ||
More Information
Set Membership: |
|

LEMD1
Genomic Map