Assay Details
Gene Symbol: NKIRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716213.3 | XP_002716259.1 | 2 - 3 | 291 | 82 | |
XM_008266059.2 | XP_008264281.1 | 2 - 3 | 244 | 82 | ||
XM_008266060.2 | XP_008264282.1 | 2 - 3 | 201 | 82 | ||
XM_017346747.1 | XP_017202236.1 | 2 - 3 | 252 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
