Assay Details
Gene Symbol: CDYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008274263.2 | XP_008272485.1 | 1153 | 58 | ||
XM_008274264.2 | XP_008272486.1 | 1062 | 58 | |||
XM_008274265.2 | XP_008272487.1 | 1047 | 58 | |||
XM_008274266.2 | XP_008272488.1 | 1036 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
