Assay Details
| Gene Symbol: LOC100344835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715453.3 | XP_002715499.1 | 5 - 6 | 767 | 100 | |
| XM_008264782.2 | XP_008263004.1 | 5 - 6 | 767 | 100 | ||
More Information
Set Membership: |
|

LOC100344835
Genomic Map