Assay Details
| Gene Symbol: LOC100349807 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263611.2 | XP_008261833.1 | 11 - 12 | 1300 | 78 | |
| XM_017344509.1 | XP_017199998.1 | 10 - 11 | 1174 | 78 | ||
| XM_017344511.1 | XP_017200000.1 | 10 - 11 | 1222 | 78 | ||
| XM_017344512.1 | XP_017200001.1 | 11 - 12 | 1302 | 78 | ||
More Information
Set Membership: |
|

LOC100349807
Genomic Map