Assay Details
| Gene Symbol: LOC100348238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262893.2 | XP_008261115.1 | 16 - 17 | 2259 | 81 | |
| XM_008262894.2 | XP_008261116.1 | 16 - 17 | 2260 | 81 | ||
| XM_008262895.2 | XP_008261117.1 | 16 - 17 | 2485 | 81 | ||
| XM_008262899.2 | XP_008261121.1 | 17 - 18 | 2317 | 81 | ||
| XM_008262901.2 | XP_008261123.1 | 17 - 18 | 2290 | 81 | ||
| XM_017345107.1 | XP_017200596.1 | 16 - 17 | 2259 | 81 | ||
| XM_017345108.1 | XP_017200597.1 | 16 - 17 | 2260 | 81 | ||
| XM_017345109.1 | XP_017200598.1 | 15 - 16 | 2067 | 81 | ||
| XM_017345110.1 | XP_017200599.1 | 16 - 17 | 2263 | 81 | ||
| XM_017345111.1 | XP_017200600.1 | 16 - 17 | 2262 | 81 | ||
| XM_017345112.1 | XP_017200601.1 | 16 - 17 | 2262 | 81 | ||
More Information
Set Membership: |
|

LOC100348238
Genomic Map