Assay Details
| Gene Symbol: LOC100348238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262893.2 | XP_008261115.1 | 17 - 18 | 2404 | 86 | |
| XM_008262894.2 | XP_008261116.1 | 17 - 18 | 2405 | 86 | ||
| XM_008262895.2 | XP_008261117.1 | 17 - 18 | 2630 | 86 | ||
| XM_008262899.2 | XP_008261121.1 | 18 - 19 | 2462 | 86 | ||
| XM_008262901.2 | XP_008261123.1 | 18 - 19 | 2435 | 86 | ||
| XM_017345107.1 | XP_017200596.1 | 17 - 18 | 2404 | 86 | ||
| XM_017345108.1 | XP_017200597.1 | 17 - 18 | 2405 | 86 | ||
| XM_017345109.1 | XP_017200598.1 | 16 - 17 | 2212 | 86 | ||
| XM_017345110.1 | XP_017200599.1 | 17 - 18 | 2408 | 86 | ||
| XM_017345111.1 | XP_017200600.1 | 17 - 18 | 2407 | 86 | ||
| XM_017345112.1 | XP_017200601.1 | 17 - 18 | 2407 | 86 | ||
More Information
Set Membership: |
|

LOC100348238
Genomic Map