Assay Details
| Gene Symbol: LOC100348238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262893.2 | XP_008261115.1 | 30 - 31 | 4611 | 67 | |
| XM_008262894.2 | XP_008261116.1 | 29 - 30 | 4486 | 67 | ||
| XM_008262895.2 | XP_008261117.1 | 30 - 31 | 4837 | 67 | ||
| XM_008262899.2 | XP_008261121.1 | 31 - 32 | 4669 | 67 | ||
| XM_008262901.2 | XP_008261123.1 | 31 - 32 | 4642 | 67 | ||
| XM_017345107.1 | XP_017200596.1 | 30 - 31 | 4608 | 67 | ||
| XM_017345108.1 | XP_017200597.1 | 30 - 31 | 4591 | 67 | ||
| XM_017345109.1 | XP_017200598.1 | 29 - 30 | 4419 | 67 | ||
More Information
Set Membership: |
|

LOC100348238
Genomic Map