Assay Details
| Gene Symbol: LOC100348238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262893.2 | XP_008261115.1 | 8 - 9 | 1122 | 61 | |
| XM_008262894.2 | XP_008261116.1 | 8 - 9 | 1123 | 61 | ||
| XM_008262895.2 | XP_008261117.1 | 8 - 9 | 1348 | 61 | ||
| XM_008262899.2 | XP_008261121.1 | 9 - 10 | 1180 | 61 | ||
| XM_008262901.2 | XP_008261123.1 | 9 - 10 | 1153 | 61 | ||
| XM_017345107.1 | XP_017200596.1 | 8 - 9 | 1122 | 61 | ||
| XM_017345108.1 | XP_017200597.1 | 8 - 9 | 1123 | 61 | ||
| XM_017345110.1 | XP_017200599.1 | 8 - 9 | 1126 | 61 | ||
| XM_017345111.1 | XP_017200600.1 | 8 - 9 | 1125 | 61 | ||
| XM_017345112.1 | XP_017200601.1 | 8 - 9 | 1125 | 61 | ||
More Information
Set Membership: |
|

LOC100348238
Genomic Map