Assay Details
| Gene Symbol: C16H1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268175.1 | XP_008266397.1 | 2 - 3 | 150 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16H1orf100| Gene Symbol: C16H1orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268175.1 | XP_008266397.1 | 2 - 3 | 150 | 79 | |
Set Membership: |
|