Assay Details
Gene Symbol: INO80E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008257939.2 | XP_008256161.1 | 3 - 4 | 447 | 75 | |
XM_008257940.2 | XP_008256162.1 | 3 - 4 | 447 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
