Assay Details
Gene Symbol: SMIM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008273713.2 | XP_008271935.1 | 106 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008273713.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
