Assay Details
| Gene Symbol: PEA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264236.2 | XP_008262458.1 | 3 - 4 | 468 | 75 | |
| XM_008264237.2 | XP_008262459.1 | 3 - 4 | 402 | 75 | ||
More Information
Set Membership: |
|

PEA15
Genomic Map