Assay Details
| Gene Symbol: RSPH10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723079.3 | XP_002723125.1 | 1113 | 49 | ||
| XM_017339080.1 | XP_017194569.1 | 1569 | 49 | |||
| XM_017339081.1 | XP_017194570.1 | 1569 | 49 | |||
More Information
Set Membership: |
|

RSPH10B
Genomic Map