Assay Details
Gene Symbol: ZNF516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008252830.2 | XP_008251052.1 | 4620 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
