Assay Details
| Gene Symbol: ADAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723773.2 | XP_002723819.1 | 1737 | 57 | ||
| XM_008252505.1 | XP_008250727.1 | 1824 | 57 | |||
| XM_008252507.1 | XP_008250729.1 | 1689 | 57 | |||
More Information
Set Membership: |
|

ADAD2
Genomic Map