Assay Details
Gene Symbol: BCL2L13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002722541.3 | XP_002722587.1 | 684 | 83 | ||
XM_008250122.2 | XP_008248344.1 | 603 | 83 | |||
XM_008250124.2 | XP_008248346.1 | 543 | 83 | |||
XM_008250125.2 | XP_008248347.1 | 496 | 83 | |||
XM_008250126.2 | XP_008248348.1 | 373 | 83 | |||
XM_008250127.2 | XP_008248349.1 | 603 | 83 | |||
XM_017338479.1 | XP_017193968.1 | 756 | 83 | |||
XM_017338481.1 | XP_017193970.1 | 607 | 83 | |||
XM_017338482.1 | XP_017193971.1 | 604 | 83 | |||
XM_017338483.1 | XP_017193972.1 | 603 | 83 | |||
XM_017338484.1 | XP_017193973.1 | 603 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
