Assay Details
Gene Symbol: TAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714675.3 | XP_002714721.2 | 5 - 6 | 527 | 66 | |
XM_008262913.2 | XP_008261135.1 | 5 - 6 | 527 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
