Assay Details
Gene Symbol: NUP35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712326.3 | XP_002712372.1 | 7 - 8 | 818 | 72 | |
XM_017342977.1 | XP_017198466.1 | 4 - 5 | 394 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
