Assay Details
| Gene Symbol: LHPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722150.3 | XP_002722196.1 | 538 | 57 | ||
| XM_017338096.1 | XP_017193585.1 | 539 | 57 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LHPP| Gene Symbol: LHPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722150.3 | XP_002722196.1 | 538 | 57 | ||
| XM_017338096.1 | XP_017193585.1 | 539 | 57 | |||
Set Membership: |
|