Assay Details
| Gene Symbol: MSANTD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261977.2 | XP_008260199.1 | 2 - 3 | 726 | 90 | |
| XM_017343615.1 | XP_017199104.1 | 2 - 3 | 723 | 90 | ||
| XM_017343616.1 | XP_017199105.1 | 2 - 3 | 864 | 90 | ||
More Information
Set Membership: |
|

MSANTD4
Genomic Map