Assay Details
| Gene Symbol: DMRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708188.2 | XP_002708234.1 | 1 - 2 | 508 | 53 | |
| XM_017341623.1 | XP_017197112.1 | 1 - 2 | 508 | 53 | ||
More Information
Set Membership: |
|

DMRT1
Genomic Map