Assay Details
Gene Symbol: RPP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713404.3 | XP_002713450.2 | 2 - 3 | 296 | 63 | |
XM_008260841.2 | XP_008259063.1 | 2 - 3 | 239 | 63 | ||
XM_008260842.2 | XP_008259064.1 | 2 - 3 | 302 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
