Assay Details
| Gene Symbol: TIMM17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272593.2 | XP_008270815.2 | 2 - 3 | 233 | 73 | |
| XM_017349730.1 | XP_017205219.1 | 2 - 3 | 256 | 73 | ||
More Information
Set Membership: |
|

TIMM17B
Genomic Map