Assay Details
Gene Symbol: LOC103351986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008273243.2 | XP_008271465.1 | 2 - 3 | 287 | 85 | |
XM_008273244.2 | XP_008271466.1 | 2 - 3 | 265 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
