Assay Details
Gene Symbol: LOC103351986 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008273243.2 | XP_008271465.1 | 4 - 5 | 711 | 89 | |
XM_008273244.2 | XP_008271466.1 | 4 - 5 | 689 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
