Assay Details
Gene Symbol: TMEM131L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267300.1 | XP_008265522.1 | 30 - 31 | 4226 | 53 | |
XM_017347287.1 | XP_017202776.1 | 30 - 31 | 4229 | 53 | ||
XM_017347288.1 | XP_017202777.1 | 30 - 31 | 4226 | 53 | ||
XM_017347289.1 | XP_017202778.1 | 30 - 31 | 4226 | 53 | ||
XM_017347290.1 | XP_017202779.1 | 30 - 31 | 4229 | 53 | ||
XM_017347291.1 | XP_017202780.1 | 30 - 31 | 4226 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
