Assay Details
Gene Symbol: LOC100345461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008248352.2 | XP_008246574.1 | 626 | 89 | ||
XM_017337618.1 | XP_017193107.1 | 626 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
