Assay Details
Gene Symbol: NSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008274075.2 | XP_008272297.1 | 3 - 4 | 449 | 66 | |
XM_017350246.1 | XP_017205735.1 | 3 - 4 | 521 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
