Assay Details
Gene Symbol: MED18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002716080.3 | XP_002716126.2 | 2 - 3 | 424 | 94 | |
XM_008265867.2 | XP_008264089.1 | 2 - 3 | 288 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
