Assay Details
| Gene Symbol: TMEM170B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262386.2 | XP_008260608.2 | 1 - 2 | 129 | 54 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM170B| Gene Symbol: TMEM170B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262386.2 | XP_008260608.2 | 1 - 2 | 129 | 54 | |
Set Membership: |
|