Assay Details
| Gene Symbol: GRPEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710299.3 | XP_002710345.1 | 2 - 3 | 405 | 83 | |
| XM_017341179.1 | XP_017196668.1 | 3 - 4 | 391 | 83 | ||
More Information
Set Membership: |
|

GRPEL2
Genomic Map