Assay Details
| Gene Symbol: NFE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256574.2 | XP_008254796.1 | 1 - 2 | 508 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008256574.2 transcripts.
Set Membership: |
|

NFE2
Genomic Map