Assay Details
| Gene Symbol: NFE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711036.3 | XP_002711082.1 | 2 - 3 | 1377 | 65 | |
| XM_008256573.2 | XP_008254795.1 | 2 - 3 | 1337 | 65 | ||
| XM_008256574.2 | XP_008254796.1 | 2 - 3 | 683 | 65 | ||
| XM_008256577.2 | XP_008254799.1 | 2 - 3 | 202 | 65 | ||
More Information
Set Membership: |
|

NFE2
Genomic Map