Assay Details
Gene Symbol: TFDP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266264.2 | XP_008264486.1 | 7 - 8 | 695 | 83 | |
XM_008266265.2 | XP_008264487.1 | 7 - 8 | 685 | 83 | ||
XM_008266266.2 | XP_008264488.1 | 7 - 8 | 933 | 83 | ||
XM_008266267.2 | XP_008264489.1 | 6 - 7 | 637 | 83 | ||
XM_008266268.2 | XP_008264490.1 | 6 - 7 | 758 | 83 | ||
XM_008266269.2 | XP_008264491.1 | 7 - 8 | 770 | 83 | ||
XM_008266270.2 | XP_008264492.1 | 9 - 10 | 850 | 83 | ||
XM_008266271.2 | XP_008264493.1 | 7 - 8 | 688 | 83 | ||
XM_008266272.2 | XP_008264494.1 | 6 - 7 | 677 | 83 | ||
XM_008266273.2 | XP_008264495.1 | 6 - 7 | 732 | 83 | ||
XM_008266274.2 | XP_008264496.1 | 7 - 8 | 689 | 83 | ||
XM_017346815.1 | XP_017202304.1 | 9 - 10 | 877 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
