Assay Details
| Gene Symbol: GZMH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269454.2 | XP_008267676.1 | 5 | 914 | 87 | |
| XM_017348231.1 | XP_017203720.1 | 5 | 1042 | 87 | ||
| XM_017348232.1 | XP_017203721.1 | 5 | 970 | 87 | ||
More Information
Set Membership: |
|

GZMH
Genomic Map