Assay Details
Gene Symbol: LOC100353768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008257490.2 | XP_008255712.1 | 2 - 3 | 822 | 70 | |
XM_008257492.2 | XP_008255714.1 | 2 - 3 | 526 | 70 | ||
XM_008257493.2 | XP_008255715.1 | 2 - 3 | 833 | 70 | ||
XM_017342308.1 | XP_017197797.1 | 2 - 3 | 814 | 70 | ||
XM_017342310.1 | XP_017197799.1 | 2 - 3 | 400 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
