Assay Details
Gene Symbol: LOC100353768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008257490.2 | XP_008255712.1 | 4 - 5 | 1076 | 65 | |
XM_008257492.2 | XP_008255714.1 | 4 - 5 | 780 | 65 | ||
XM_008257493.2 | XP_008255715.1 | 4 - 5 | 1087 | 65 | ||
XM_017342308.1 | XP_017197797.1 | 4 - 5 | 1068 | 65 | ||
XM_017342309.1 | XP_017197798.1 | 3 - 4 | 475 | 65 | ||
XM_017342310.1 | XP_017197799.1 | 4 - 5 | 654 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
