Assay Details
| Gene Symbol: TMEM207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266652.2 | XP_008264874.1 | 4 - 5 | 438 | 81 | |
| XM_008266654.2 | XP_008264876.1 | 3 - 4 | 393 | 81 | ||
| XM_017346987.1 | XP_017202476.1 | 3 - 4 | 319 | 81 | ||
More Information
Set Membership: |
|

TMEM207
Genomic Map