Assay Details
Gene Symbol: SYNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249555.2 | XP_008247777.2 | 621 | 61 | ||
XM_008249556.2 | XP_008247778.2 | 621 | 61 | |||
XM_017338163.1 | XP_017193652.1 | 621 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
