Assay Details
| Gene Symbol: LOC100350827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262483.2 | XP_008260705.1 | 3 - 4 | 431 | 94 | |
| XM_017344959.1 | XP_017200448.1 | 4 - 5 | 535 | 94 | ||
More Information
Set Membership: |
|

LOC100350827
Genomic Map