Assay Details
| Gene Symbol: PPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714299.3 | XP_002714345.1 | 5 - 6 | 1053 | 67 | |
| XM_008262617.2 | XP_008260839.1 | 5 - 6 | 698 | 67 | ||
| XM_008262618.2 | XP_008260840.1 | 5 - 6 | 611 | 67 | ||
| XM_008262619.2 | XP_008260841.1 | 5 - 6 | 653 | 67 | ||
More Information
Set Membership: |
|

PPT2
Genomic Map