Assay Details
| Gene Symbol: PHETA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272236.2 | XP_008270458.2 | 371 | 79 | ||
| XM_017349624.1 | XP_017205113.1 | 377 | 79 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008272236.2, XM_017349624.1 transcripts.
Set Membership: |
|

PHETA1
Genomic Map