Assay Details
Gene Symbol: KIRREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264256.2 | XP_008262478.1 | 5 - 6 | 770 | 94 | |
XM_008264257.2 | XP_008262479.1 | 5 - 6 | 770 | 94 | ||
XM_008264258.2 | XP_008262480.1 | 4 - 5 | 620 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
