Assay Details
| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264256.2 | XP_008262478.1 | 8 - 9 | 1150 | 84 | |
| XM_008264258.2 | XP_008262480.1 | 7 - 8 | 1000 | 84 | ||
More Information
Set Membership: |
|

KIRREL1
Genomic Map