Assay Details
Gene Symbol: KIRREL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264256.2 | XP_008262478.1 | 9 - 10 | 1325 | 55 | |
XM_008264257.2 | XP_008262479.1 | 9 - 10 | 1277 | 55 | ||
XM_008264258.2 | XP_008262480.1 | 8 - 9 | 1175 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
